Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leucine, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603786 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

L1202, 61-90-5, C6H13NO2

Spectrum™ Chemical Leucine, USP is a hydrophobic essential amino acid that is a component in astacin, ferritin and other buffer proteins. It is used for the creation of sterols in the muscle and adipose tissues. Widely used in sport supplements for its muscle building properties, it is also used as a food additive and flavor enhancer. Spectrum Chemical manufactured USP LEUCINE products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

CAS Max % 100%
Chloride 0.05%
Ignition Residue 0.4%
Loss on Drying 0.2%
Packaging Poly Pail
Specific Rotation +14.9° to +17.3°
Sulfate 0.03%
Quantity 5 kg
Percent Purity 98.5 to 101.5%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.