Learn More
Greiner Bio-One Leucosep Polypropylene Centrifugation Tubes
GREENER_CHOICE
Catalog No. p-7075348 Shop All Greiner Bio-One Products
Click to view available options
Capacity (Metric):
12 mL
50 mL
Sterility:
Aseptically Manufactured
Sterile
Bottom Shape:
Conical
Round
For efficient separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow.
  • Enrichment directly from whole blood
  • Simplified filling through porous barrier
  • Rapid separation in 15 minutes at room temperature
  • Removal of erythrocytes and granulocytes
  • No recontamination with erythrocytes
  • Service Plan
    No
    TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 12 mL
Material Polypropylene
For Use With Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow
Includes Porous barrier
Sterility Aseptically Manufactured
Bottom Shape Round
Product Type Centrifuge Tube (Pre-filled)
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.