Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leucovorin Calcium, USP, 95-105%, Spectrum™ Chemical
SDP

Catalog No. 18603793 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-793 100 g Amber Glass Bottle
18-603-788 100 mg Amber Glass Bottle
18-603-789 500 mg Amber Glass Bottle
18-603-790 1 g Amber Glass Bottle
18-603-791 5 g Amber Glass Bottle
18-603-792 25 g Amber Glass Bottle
18-603-794 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-603-793 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. LE117100GMBL
Only null left
Add to Cart
Add to Cart
Request a Quote

LE117, 1492-18-8, C20H21CaN7O7

Spectrum™ Chemical Leucovorin Calcium, USP is used to treat or prevent serious blood cell disorders caused by certain drugs that can reduce the effects of folic acid in the body. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1492-18-8
Molecular Formula C20H21CaN7O7
Molecular Weight (g/mol) 511.51
MDL Number MFCD00006704
InChI Key KVUAALJSMIVURS-AEQORCIRNA-L
IUPAC Name calcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate
SMILES [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 95 to 105%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.