Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leucyl-cystinyl Aminopeptidase/LNPEP Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290022 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290022 Supplier Novus Biologicals Supplier No. NBP232111PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Leucyl-cystinyl Aminopeptidase/LNPEP Polyclonal antibody specifically detects Leucyl-cystinyl Aminopeptidase/LNPEP in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Leucyl-cystinyl Aminopeptidase/LNPEP
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias angiotensin IV receptor, AT (4) receptor, CAP, Cystinyl aminopeptidase, EC 3.4.11, EC 3.4.11.3, Insulin-regulated membrane aminopeptidase, Insulin-responsive aminopeptidase, IRAP, leucyl/cystinyl aminopeptidase, leucyl-cystinyl aminopeptidase, otase, Oxytocinase, Placental leucine aminopeptidase, PLAP, P-LAPinsulin-regulated aminopeptidase, vasopressinase
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Oxytocinase using the numbering given in entry NP_005566.2 (GeneID 4012).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 4012
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.