Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukotriene A4 Hydrolase/LTA4H Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB082983 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB082983 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB082983 Supplier Novus Biologicals Supplier No. NBP299029F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Leukotriene A4 Hydrolase/LTA4H Polyclonal specifically detects Leukotriene A4 Hydrolase/LTA4H in Human samples. It is validated for ELISA.

Specifications

Antigen Leukotriene A4 Hydrolase/LTA4H
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Leukotriene A4 Hydrolase/LTA4H (NP_000886.1; Met 1-Asp 611).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4048
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.