Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukotriene A4 Hydrolase/LTA4H Antibody (OTI1E9), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP271189IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271189IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271189IR Supplier Novus Biologicals Supplier No. NBP271189IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Leukotriene A4 Hydrolase/LTA4H Monoclonal specifically detects Leukotriene A4 Hydrolase/LTA4H in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Leukotriene A4 Hydrolase/LTA4H
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI1E9
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.3.2.6, Leukotriene A(4) hydrolase, leukotriene A4 hydrolase, leukotriene A-4 hydrolase, LTA4, LTA-4 hydrolase
Gene Symbols LTA4H
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human Leukotriene A4 Hydrolase/LTA4H expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 4048.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.