Learn More
Description
Specifications
Specifications
| Antigen | Leukotriene B4 R1 |
| Applications | Flow Cytometry, Immunoassay, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | 202/7B1 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay |
| Gene Accession No. | Q15722 |
| Gene Alias | chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 |
| Gene Symbols | LTB4R |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
