Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Leukotriene B4 R1 Antibody (202/7B1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
| Antigen | Leukotriene B4 R1 |
|---|---|
| Clone | 202/7B1 |
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay |
| Applications | Immunofluorescence, Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry, Flow Cytometry |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017653
![]() |
Novus Biologicals
NB100648310.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10064831
![]() |
Novus Biologicals
NB10064831 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Leukotriene B4 R1 Monoclonal specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoassay.Specifications
| Leukotriene B4 R1 | |
| 1.0 mg/mL | |
| Immunofluorescence, Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry, Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| GPCR, Immunology, Innate Immunity, Signal Transduction | |
| Q15722 | |
| 1241 | |
| HeLa cell transfectants expressing human BLTR. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 202/7B1 | |
| Flow Cytometry 1:100-1:200, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Immunoassay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
| LTB4R | |
| IgG2a | |
| Protein G purified | |
| NB100-64831 recognizes the human BLTR molecule, the leukotriene B4 receptor. BLTR is expressed by peripheral blood granulocytes and is important in the pro-inflammatory response. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title