Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukotriene B4 R1 Antibody (202/7B1), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB164831IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164831IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164831IR Supplier Novus Biologicals Supplier No. NB10064831IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Leukotriene B4 R1 Monoclonal specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow Cytometry, Immunoassay, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen).

Specifications

Antigen Leukotriene B4 R1
Applications Flow Cytometry, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 202/7B1
Conjugate DyLight 755
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7
Gene Symbols LTB4R
Host Species Mouse
Immunogen HeLa cell transfectants expressing human BLTR.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1241
Test Specificity NB100-64831 recognizes the human BLTR molecule, the leukotriene B4 receptor. BLTR is expressed by peripheral blood granulocytes and is important in the pro-inflammatory response.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.