Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukotriene B4 R1 Antibody (203/14F11), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB174675 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174675 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174675 Supplier Novus Biologicals Supplier No. FAB099I
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Leukotriene B4 R1 Monoclonal antibody specifically detects Leukotriene B4 R1 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Leukotriene B4 R1
Applications Flow Cytometry
Classification Monoclonal
Clone 203/14F11
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7
Host Species Mouse
Immunogen HeLa cervical epithelial carcinoma cell line transfected with human Leukotriene B4 R1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1241
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.