Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Leukotriene B4 R1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227422SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227422SS 0.025 mg
NBP227422 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227422SS Supplier Novus Biologicals Supplier No. NBP227422SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

Leukotriene B4 R1 Polyclonal specifically detects Leukotriene B4 R1 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Leukotriene B4 R1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5ug/ml∼, Flow Cytometry reported in scientific literature (PMID 28763622), Immunohistochemistry, Immunohistochemistry-Paraffin 10ug/ml∼
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q15722
Gene Alias chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7
Gene Symbols LTB4R
Host Species Rabbit
Immunogen A portion of amino acids 1-60 from human BLT1 was used as the immunogen for this antibody (1st extracellular domain).
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1241
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and white-cheeked gibbon, 94% in common chimpanzee, 83% in orangutan, and 78% in cow, new world monkey, and rhesus monkey.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.