Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Leukotriene B4
GREENER_CHOICE

Catalog No. 230750u
Click to view available options
Quantity:
50 μg
This item is not returnable. View return policy
This item is not returnable. View return policy

BLT1/BLT2 receptor agonist and potent chemotactic factor

Potent lipid inflammatory mediator derived from the 5-lipoxygenase pathway of arachidonic acid metabolism. Binds to BLT1 and BLT2 receptors and acts as a potent chemotactic agent and activator of leukocytes. Also displays antiviral activity towards DNA viruses and retroviruses.

Chemical Identifiers

CAS 71160-24-2
Molecular Formula C20H32O4

Specifications

Quantity 50 μg
Formula Weight 336.47
Purity 97%
Molecular Weight (g/mol) 336.472
Chemical Name or Material Leukotriene B4
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.