Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Levcromakalim
GREENER_CHOICE

Catalog No. 137850
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Kir6 (KATP) channel opener; active enantiomer of cromakalim (Cat. No. 1377)

Levcromakalim is an active enantiomer of the prototypical Kir6 (KATP) channel opener Cromakalim. Hypotensive and airways relaxant. IC50 = 490 nM in guinea pig trachea.

Chemical Identifiers

CAS 94535-50-9
Molecular Formula C16H18N2O3

Specifications

Quantity 50 mg
Formula Weight 286.33
Purity >99%
Molecular Weight (g/mol) 286.331
Chemical Name or Material Levcromakalim
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.