Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Levcromakalim
GREENER_CHOICESDP

Catalog No. 137810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-781-0 10 mg
13-785-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 13-781-0 Supplier Tocris Bioscience™ Supplier No. 1378/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Kir6 (KATP) channel opener; active enantiomer of cromakalim (Cat. No. 1377)

Levcromakalim is an active enantiomer of the prototypical Kir6 (KATP) channel opener Cromakalim. Hypotensive and airways relaxant. IC50 = 490 nM in guinea pig trachea.

Chemical Identifiers

CAS 94535-50-9
Molecular Formula C16H18N2O3

Specifications

Quantity 10 mg
Formula Weight 286.33
Purity >99%
Molecular Weight (g/mol) 286.331
Chemical Name or Material Levcromakalim
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.