Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Levetiracetam
GREENER_CHOICE

Catalog No. p-4784106
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Antiepileptic; binds SV2A

Levetiracetam is an antiepileptic that displays distinctive properties from conventional antiepileptic drugs. Displays potent seizure protection in animal models of chronic epilepsy but lacks activity in acute seizure models. Binds synaptic vesicle protein 2A (SV2A) and inhibits Na+-dependent Cl-/HCO3- exchange. In a mouse model of Alzheimer's disease, Levetiracetam rescues levels of presynaptic vesicle fusion and neurotransmitter release, reduces Aβ42 and APP levels and decreases deposits of amyloid plaque; in vitro Levetiracetam inhibits Aβ-induced glutamate from astrocytes.

Chemical Identifiers

CAS 102767-28-2
Molecular Formula C8H14N2O2

Specifications

Quantity 10 mg
Formula Weight 170.21
Purity >99%
Molecular Weight (g/mol) 170.21
Chemical Name or Material Levetiracetam
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.