Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Levodopa, Powder, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603810 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
10 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-810 500 g Amber Glass Bottle
18-603-807 5 g Amber Glass Bottle
18-603-808 25 g Amber Glass Bottle
18-603-809 100 g Amber Glass Bottle
18-603-812 10 kg Poly Pail
18-603-811 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-603-810 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. D1066500GM
Only null left
Add to Cart
Add to Cart

D1066, 59-92-7, C9H11NO4

Spectrum™ Chemical Levodopa, Powder, USP is used to treat the stiffness, tremors, spasms, and poor muscle control of Parkinson′s disease. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 59-92-7
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00002598
InChI Key WTDRDQBEARUVNC-LURJTMIESA-N
IUPAC Name (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation -160° to -167°
Percent Purity 98 to 102%
Grade USP
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.