Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lewis A,B Blood Group Antigens Antibody (HEA164), PE, Novus Biologicals™
SDP

Catalog No. NB154089 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB154089 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB154089 Supplier Novus Biologicals Supplier No. NBP312056PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Lewis A,B Blood Group Antigens Monoclonal antibody specifically detects Lewis A,B Blood Group Antigens in Human samples. It is validated for Flow Cytometry

Specifications

Antigen Lewis A,B Blood Group Antigens
Applications Flow Cytometry
Classification Monoclonal
Clone HEA164
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias Le-a, Le-b, Lewis A, Lewis A Blood Group Antigen, Lewis B, lewis B blood group antigen
Host Species Rabbit
Immunogen This antibody was raised by immunising mice with human carcinoma cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.