Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lewis A Blood Group Antigen Antibody (7LE) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB500525 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500525 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500525 Supplier Novus Biologicals Supplier No. NB500525
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Lewis A Blood Group Antigen Monoclonal specifically detects Lewis A Blood Group Antigen in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Lewis A Blood Group Antigen
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7LE
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:2000, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Gene Accession No. P21217
Gene Alias A transferase, A3GALNT, A3GALT1, ABO, B transferase, EC=2.4.1.37, EC=2.4.1.40, Fucosylglycoprotein 3-alpha-galactosyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase, Fucosylglycoprotein alpha-N-acetylgalactosaminyltransferase soluble form, Glycoprotein-fucosylgalactoside alpha-galactosyltransferase, Glycoprotein-fucosylgalactoside alpha-N-acetylgalactosaminyltransferase, GTB, Histo-blood group A transferase, Histo-blood group ABO system transferase, Histo-blood group B transferase, NAGAT
Gene Symbols ABO
Host Species Mouse
Immunogen Mucin isolated by size-exclusion chromatography from an ovarian cyst fluid (pure endocervical type according to the Fenoglio's classification).
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2525
Test Specificity The antibody 7LE recognizes Lewisa blood group antigen, a carbohydrate determinant carried on both glycolipids and glycoproteins, expressed in colonic epithelial cells. Lewisa may be useful for detection of gastrointestinal, pancreatic and colorectal tumors.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.