Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lewis Y Antibody (BR96), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB015504 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB015504 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB015504 Supplier Novus Biologicals Supplier No. NBP281077AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Lewis Y Monoclonal specifically detects Lewis Y in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lewis Y
Applications ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone BR96
Conjugate Alexa Fluor 488
Dilution ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias IPCA-2, IPCA6, IPCA-6, IPCA-8, PCANAP2, PCANAP6, PCANAP8, Prostate cancer-associated protein 6, PRST, Solute carrier family 45 member 3
Host Species Rabbit
Immunogen H3396 cultured line established from a metastasis of a human breast adenocarcinoma.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody binds Lewis Y with a Ka of 0.1 uM.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.