Learn More
Invitrogen™ LFNG Monoclonal Antibody (4F8)

Description
Peptide Sequence: MTPGRCCLAA DIQVETFIFT DGEDEALARH TGNVVITNCS AAHSRQALSC KMAVEYDRFI ESGRKWFCHV DDDNYVNLRA LLRLLASYPH TRDVYVGKPS LDRPIQAMER VSENKVRPVH FWFATGGAGF CISRGLALKM SPWASGGHFM NTAERIRLPD DCTIGYIVEA LLGVPLIRSG LFHSHLENLQ QVPTSELHEQ VTLSYGMFEN KRNAVHVKGP FSVEADPSRF RSIHCHLYPD TPWCPRTAIF.
Specifications
Specifications
| Antigen | LFNG |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | 4F8 |
| Concentration | 0.2-1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | LFNG |
| Gene Accession No. | Q8NES3 |
| Gene Alias | AW061165; beta-1,3-N-acetylglucosaminyltransferase lunatic fringe; LFNG; LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; lunatic fringe; lunatic fringe gene homolog; MGC128664 protein; O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase; SCDO3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.