Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgi1 Antibody (S283-7), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB242190488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB242190488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB242190488 Supplier Novus Biologicals Supplier No. NBP242190AF488
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lgi1 Monoclonal specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Lgi1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S283-7
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1
Gene Symbols LGI1
Host Species Mouse
Immunogen Fusion protein amino acids 37-113 (LRRNT domain and first LRR repeat) of mouse LGI1. Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ∼50% identity with LGI2, LGI3 and LGI4.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9211
Test Specificity approx 50% identity with LGI2, LGI3 and LGI4.Detects approx 60kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.