Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LGP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110841
Description
Immunogen sequence: GLLTNEISMV QARGRARADQ SVYAFVATEG SRELKRELIN EALETLMEQA VAAVQKMDQA EYQAKIRDLQ QAALTKRAAQ AAQRENQRQQ FPVEHVQLLC INCMVAV.
Anti-viral innate immune responses are triggered by pathogen-associated molecular patters (such as the accumulation of intracellular nucleic acids resulting from virus infections) and represent the first line of defense against numerous infectious organisms. The Toll-like receptors (TLR) 3, 7, 8, and 9 are expressed in immune cells and function as transmembrane pattern recognition receptors to detect foreign nucleic acids. Other proteins that play similar roles, such as RIG-1 and MDA5 are members of a CARD-helicase family and are expressed in the cytoplasm. A third protein, LGP2, is similar to RIG-1 and MDA5, except for lacking a homologous CARD domain. It is thought to act as an element of negative-feedback regulation of intracellular antiviral signaling. At least four isoforms of LGP2 are known to exist.
Specifications
LGP2 | |
Polyclonal | |
Unconjugated | |
DHX58 | |
B430001I08Rik; D11LGP2; D11lgp2e; DEXH (Asp-Glu-X-His) box polypeptide 58; DEXH-box helicase 58; DHX58; Lgp2; LPG2; ortholog of mouse D11lgp2; probable ATP-dependent helicase LGP2; Probable ATP-dependent RNA helicase DHX58; protein D11Lgp2; Protein D11Lgp2 homolog; RGD1310093; RIG-I-like receptor 3; RIG-I-like receptor Lgp2; RLR; RLR-3; RNA helicase LGP2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79132 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96C10 | |
DHX58 | |
Recombinant Protein corresonding to Human LGP2. Recombinant protein control fragment (Product #RP-92404). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction