Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr4/GPR48 Antibody (852229), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB374144 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374144 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374144 Supplier Novus Biologicals Supplier No. FAB7750E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lgr4/GPR48 Monoclonal antibody specifically detects Lgr4/GPR48 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF

Specifications

Antigen Lgr4/GPR48
Applications Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 852229
Conjugate DyLight 405
Dilution Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias GPR48G protein-coupled receptor 48, G-protein coupled receptor 48, leucine-rich repeat containing G protein-coupled receptor 4
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Lgr4/GPR48, Met1-Asp951, Accession # Q9BXB1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 55366
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.