Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr5/GPR49 Antibody (707042), HRP, Novus Biologicals™
SDP

Catalog No. NB346337 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346337 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346337 Supplier Novus Biologicals Supplier No. FAB8078H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Human samples. It is validated for Agonist Activity, CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Lgr5/GPR49
Applications Agonist Activity, CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 707042
Conjugate HRP
Dilution Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready
Formulation PBS
Gene Alias FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Lgr5/GPR49, Met1-Ile560, Accession # O75473
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8549
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.