Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr5/GPR49 Antibody (803420), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB346239 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346239 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346239 Supplier Novus Biologicals Supplier No. FAB8240MFV500
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Mouse samples. It is validated for Agonist Activity, CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Lgr5/GPR49
Applications Agonist Activity, CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 803420
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry, Agonist Activity, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Host Species Rat
Immunogen E. coli-derived recombinant mouse Lgr5/GPR49, Lys375-Pro547, Accession # Q9Z1P4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8549
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.