Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr5/GPR49 Antibody (CL13579), Novus Biologicals™
SDP

Catalog No. NB306027 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306027 25 μL
NB306028 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB306027 Supplier Novus Biologicals Supplier No. NBP32457125UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lgr5/GPR49 Monoclonal antibody specifically detects Lgr5/GPR49 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Lgr5/GPR49
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13579
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38
Host Species Mouse
Immunogen This antibody was designed to target the recombinant protein Lgr5/GPR49, with the precise immunogen sequence being withheld for proprietary reasons.
Purification Method Protein A purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 8549
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.