Learn More
Description
Specifications
Specifications
| Antigen | Lgr5/GPR49 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot 1.0-2.0 μg/mL, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen |
| Formulation | PBS with No Preservative |
| Gene Accession No. | O75473 |
| Gene Alias | FEX, G protein-coupled receptor 49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 |
| Gene Symbols | LGR5 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
