Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Lgr5/GPR49 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB8240R025UG
This item is not returnable.
View return policy
Description
Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Mouse samples. It is validated for Flow Cytometry.Specifications
| Lgr5/GPR49 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.1 ug/10^6 cells | |
| Q9Z1P4 | |
| LGR5 | |
| E. coli-derived recombinant mouse Lgr5/GPR49 Lys375-Pro547 Accession # Q9Z1P4 | |
| 25 μg | |
| Primary | |
| Detects mouse Lgr5/GPR49 in direct ELISAs. Stains mouse Lgr5 transfected cells but not irrelevant transfectants in flow cytometry. This clone, also known as “RD20”, has been found to detect an epitope in C-terminal LRR cap of LGR5 (Ref. 1). | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| Flow Cytometry | |
| 803420 | |
| Alexa Fluor 647 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8549 | |
| Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction