Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP14969505 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP14969505 0.05 mg
NB018654 0.025 mg
NBP149695 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP14969505 Supplier Novus Biologicals Supplier No. NBP1496950.05MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lgr6 Polyclonal specifically detects Lgr6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Lgr6
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA < 1:10000
Formulation Tris/Glycine buffer pH 7.5-7.9, stabilizing protein, and glycerol with 0.02% sodium azide
Gene Accession No. Q9HBX8
Gene Alias FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631
Gene Symbols LGR6
Host Species Rabbit
Immunogen Synthetic peptides corresponding to unique epitope on Lgr6. The peptide sequence corresponding to amino acids 860-915 of the human Lgr6 protein, the peptide was post-synthetically modified to achieve desired antigenicity before coupling to a carrier protein (CAI15929).
Quantity 0.05 mg
Primary or Secondary Primary
Gene ID (Entrez) 59352
Test Specificity This antibody detects a fuzzy band of approximately 115 kDa in PC-Lgr6 samples and few additional bands between MW45-60kDa were also observed. The identity of these bands are not known but could be proteolytic fragment of the full-length Lgr6 molecule.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.