Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lgr6 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$243.50 - $670.50

Specifications

Antigen Lgr6
Dilution Western Blot 1:100-1:2000, ELISA < 1:10000
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB018654
SDP
View Documents
Novus Biologicals
NBP1496950.025MG
0.025 mg
Each for $243.50
Only null left
Add to Cart
 
NBP14969505
SDP
View Documents
Novus Biologicals
NBP1496950.05MG
0.05 mg
Each for $402.00
Only null left
Add to Cart
 
NBP149695
SDP
View Documents
Novus Biologicals
NBP149695
0.1 mg
Each for $670.50
Only null left
Add to Cart
 
Description

Description

Lgr6 Polyclonal specifically detects Lgr6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Lgr6
Western Blot, ELISA
Unconjugated
Mouse, Rat, Human
FLJ14471, gonadotropin receptor, GPCR, leucine-rich repeat containing G protein-coupled receptor 6, leucine-rich repeat-containing G protein-coupled receptor 6, leucine-rich repeat-containing G-protein coupled receptor 6, VTS20631
LGR6
IgG
This antibody detects a fuzzy band of approximately 115 kDa in PC-Lgr6 samples and few additional bands between MW45-60kDa were also observed. The identity of these bands are not known but could be proteolytic fragment of the full-length Lgr6 molecule.
Western Blot 1:100-1:2000, ELISA < 1:10000
Polyclonal
Rabbit
Q9HBX8
59352
Synthetic peptides corresponding to unique epitope on Lgr6. The peptide sequence corresponding to amino acids 860-915 of the human Lgr6 protein, the peptide was post-synthetically modified to achieve desired antigenicity before coupling to a carrier protein (CAI15929).
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.