Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LH beta Antibody (L1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB10073010N Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10073010N 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10073010N Supplier Novus Biologicals Supplier No. NB10073010AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LH beta Monoclonal specifically detects LH beta in Human samples. It is validated for ELISA, Sandwich ELISA, Western Blot.

Specifications

Antigen LH beta
Applications ELISA, Sandwich ELISA, Western Blot
Classification Monoclonal
Clone L1
Conjugate Alexa Fluor 700
Dilution Western Blot, Sandwich ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CGB4, hLHB, interstitial cell stimulating hormone, beta chain, LSH-B, LSH-beta, luteinizing hormone beta polypeptide, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta
Gene Symbols LHB
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified natural LH beta.
Purification Method Protein A or G purified
Quantity 0.2 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3972
Test Specificity High affinity for the beta chain of LH. No cross-reaction with FSH, TSH or HCG.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.