Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ Licarbazepine

Click to view available options
Quantity:
10 mg
50 mg
Description
Licarbazepine is an active metabolite of oxcarbazepine (Cat. No. 3864). Produces dose-dependent inhibition of glutamatergic excitatory postsynaptic potentials (EPSPs). Displays anticonvulsant activity; exhibits minor potentiation of GABAA receptor currents.
Specifications
Specifications
Quantity | 10 mg |
Product Type | Licarbazepine |
Purity | 0.98 |
Molecular Weight (g/mol) | 254.28 |
Inhibitors | Licarbazepine |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction