Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 125-3
Catalog No. M1501757184 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
DIOL5
NH2
No Phase
RP18
RP8
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase
M1501757184 No Phase
M1501757075 DIOL5
M1501757076 NH2
M1501757092 RP8
M1501757127 RP18
M1501757174 No Phase
M1501757175 No Phase
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. M1501757184 Supplier MilliporeSigma™ Supplier No. 1.50175.7184
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma LiChroCART™ HPLC Cartridge, 125-3 comes with customized packing (please specify packing material) and is 125mmL x 3mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 3 mm
Length (Metric) 125 mm
Stationary Phase No Phase
Packing Material Customized (specify packing material)
Column Type LiChroCART™ HPLC Cartridge