Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 55-4
Catalog No. M1502287078 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
HPLC phase
RP18
RP8
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase
M1502287078 HPLC phase
M1502287139 RP8
M1502287085 RP18
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1502287078 Supplier MilliporeSigma™ Supplier No. 1502287078
Only null left
Add to Cart
Add to Cart
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma LiChroCART™ HPLC Cartridge, 55-4 comes with customized packing (please specify packing material) and is 55mmL x 4mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4 mm
Length (Metric) 55 mm
Mobile Phase Compatibility RP (reversed-phase)
Stationary Phase HPLC phase
Packing Material Customized (specify packing material)
Column Type LiChroCART™ HPLC Cartridge