Learn More
MilliporeSigma™ LiChroCART™ HPLC Cartridge, 75-4
Catalog No. M1501717131 Shop All MilliporeSigma Products
Change view
Click to view available options
Stationary Phase:
60 SI
CN5
RP18
RP8
Mobile Phase Compatibility:
NP (normal-phase)
RP (reversed-phase)
RP (when in mode) and NP
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Stationary Phase Mobile Phase Compatibility
M1501717131 RP18 RP (reversed-phase)
M1501717071 CN5 RP (when in mode) and NP
M1501717109 60 SI NP (normal-phase)
M1501717194 RP8 RP (reversed-phase)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1501717131 Supplier MilliporeSigma™ Supplier No. 1501717131
Available in a variety of stationary phases that function in normal and/or reverse phase. MilliporeSigma LiChroCART™ HPLC Cartridge, 75-4 comes with customized packing (please specify packing material) and is 75mmL x 4mmI.D.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 4 mm
Length (Metric) 75 mm
Mobile Phase Compatibility RP (reversed-phase)
Stationary Phase RP18
Packing Material Customized (specify packing material)
Column Type LiChroCART™ HPLC Cartridge