Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIF Antibody (39N7D10), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP227406A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP227406A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP227406A94 Supplier Novus Biologicals Supplier No. NBP227406AF594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LIF Monoclonal specifically detects LIF in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen LIF
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 39N7D10
Conjugate Alexa Fluor 594
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Gene Symbols LIF
Host Species Rat
Immunogen A recombinant murine Lif protein containing amino acids 24-203 was used as the immunogen for this antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines
Primary or Secondary Primary
Gene ID (Entrez) 3976.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.