Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIF Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB206709 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB206709 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB206709 Supplier Novus Biologicals Supplier No. NBP176554B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIF Polyclonal antibody specifically detects LIF in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen LIF
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the center of human LIF. The immunogen is located within amino acids 50 - 100 of LIF. Amino Acid Squence: LYYTAQGEPFPNNLDK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines
Primary or Secondary Primary
Gene ID (Entrez) 3976
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.