Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIF Antibody (MM0454-6H31), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB361722 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB361722 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB361722 Supplier Novus Biologicals Supplier No. NBP211773MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIF Monoclonal antibody specifically detects LIF in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization

Specifications

Antigen LIF
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Neutralization
Classification Monoclonal
Clone MM0454-6H31
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias CDF, D factor, DIA, differentiation inhibitory activity, differentiation stimulating factor, Differentiation-stimulating factor, Emfilermin, HILDAcholinergic differentiation factor, leukemia inhibitory factor, leukemia inhibitory factor (cholinergic differentiation factor), Melanoma-derived LPL inhibitor, MLPLI
Host Species Mouse
Immunogen Human recombinant LIF
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Stem Cell Lines
Primary or Secondary Primary
Gene ID (Entrez) 3976
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.