Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIFR alpha Antibody (110), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB075228 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075228 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075228 Supplier Novus Biologicals Supplier No. NBP290580AF750
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LIFR alpha Monoclonal specifically detects LIFR alpha in Mouse samples. It is validated for ELISA.

Specifications

Antigen LIFR alpha
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate Alexa Fluor 750
Formulation 50 mM Sodium Borate
Gene Alias CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse LIFR alpha (NP_038612.1; Met 1-Ser 828).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3977
Test Specificity No cross-reactivity in ELISA with: Human LIFR
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.