Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIFR alpha Antibody (110), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB332682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB332682 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB332682 Supplier Novus Biologicals Supplier No. NBP290580MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LIFR alpha Monoclonal antibody specifically detects LIFR alpha in Mouse samples. It is validated for ELISA

Specifications

Antigen LIFR alpha
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate mFluor Violet 450 SE
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Mouse LIFR alpha (Accession#: NP_038612.1, Met 1-Ser 828)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3977
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.