Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIFR alpha Antibody (32953), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB175549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175549 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175549 Supplier Novus Biologicals Supplier No. FAB249K
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

LIFR alpha Monoclonal antibody specifically detects LIFR alpha in Human samples. It is validated for Western Blot, Flow Cytometry

Specifications

Antigen LIFR alpha
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 32953
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human LIF R alpha , Gln45-Ser833, Accession # P42702
Purification Method Protein A or G purified from ascites
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3977
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.