Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIFR alpha Antibody (673602), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB311842 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311842 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311842 Supplier Novus Biologicals Supplier No. FAB5990PECY55
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LIFR alpha Monoclonal specifically detects LIFR alpha in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen LIFR alpha
Applications Flow Cytometry
Classification Monoclonal
Clone 673602
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LIF R alpha , Leu44-Ser828, Accession # P42703
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3977
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.