Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIFR alpha Antibody (MM0455-9B23), HRP, Novus Biologicals™
SDP

Catalog No. NBP211774H Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211774H 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211774H Supplier Novus Biologicals Supplier No. NBP211774H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIFR alpha Monoclonal specifically detects LIFR alpha in Human samples. It is validated for Western Blot.

Specifications

Antigen LIFR alpha
Applications Western Blot
Classification Monoclonal
Clone MM0455-9B23
Conjugate HRP
Dilution Western Blot
Formulation PBS with No Preservative
Gene Alias CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Gene Symbols LIFR
Host Species Mouse
Immunogen Human recombinant LIFR alpha (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3977
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.