Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIFR alpha Rabbit anti-Mouse, Clone: 110, Novus Biologicals™
SDP

Catalog No. NB024907 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB024907 100 μg
NB024908 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB024907 Supplier Novus Biologicals Supplier No. NBP290580100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

LIFR alpha Monoclonal specifically detects LIFR alpha in Mouse samples. It is validated for ELISA.

Specifications

Antigen LIFR alpha
Applications ELISA
Classification Monoclonal
Clone 110
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Host Species Rabbit
Immunogen Recombinant Mouse LIFR alpha protein
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3977
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.