Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIGHT/TNFSF14 Antibody (885310), PerCP, Novus Biologicals™
SDP

Catalog No. NB367606 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB367606 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB367606 Supplier Novus Biologicals Supplier No. FAB17942C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

LIGHT/TNFSF14 Monoclonal antibody specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen LIGHT/TNFSF14
Applications Flow Cytometry
Classification Monoclonal
Clone 885310
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse LIGHT/TNFSF14, Asp72-Val239, Accession # Q9QYH9
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8740
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.