Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIGHT/TNFSF14 Antibody (MM0456-8C34), DyLight 488, Novus Biologicals™
SDP

Catalog No. NBP211964G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP211964G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP211964G Supplier Novus Biologicals Supplier No. NBP211964G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen LIGHT/TNFSF14
Applications Flow Cytometry
Classification Monoclonal
Clone MM0456-8C34
Conjugate DyLight 488
Dilution Flow Cytometry
Gene Accession No. O43557
Gene Alias CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Gene Symbols TNFSF14
Host Species Mouse
Immunogen Human recombinant LIGHT/TNFSF14 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 8740
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.