Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Lightning-Link (R) APC-Cy7 Antibody Labeling Kit
SDP

Catalog No. p-7070051 Shop All R&D Systems Products
Click to view available options
Quantity:
1 mg
1 x 100 μg Reaction
3 x 0.01 mg Reaction
300 μg

Applications: Flow Cytometry

APC/Cy7 is a tandem conjugate. In the Lightning-Link APC/Cy7 the tandem is excited at 633nm, wavelength at which the APC has its maximum absorption. The excited APC functions then as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the APC in the form of long wavelength light at 776nm.

Order Info

Quantity

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 1 x 100 μg Reaction
Conjugate APC/Cy7
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent
Product Type APC-Cy7 Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.