Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) PE-Cy7 Antibody Labeling Kit
Shop All R&D Systems Products

Click to view available options
Quantity:
180 μg
60 μg
600 μg
Description
PE/Cy7 is a tandem conjugate. In our Lightning-Link PE/Cy7 the best result is obtained when the tandem is excited at 535nm, wavelength at which the PE has its maximum absorption. The excited PE functions as an energy donor, where the energy is transferred via FRET (Fluorescence Resonance Energy Transfer) to the Cy7. The Cy7 emits then the energy received from the PE in the form of long wavelength light at 776nm.
Order Info
Quantity
Specifications
Specifications
Quantity | 60 μg |
Conjugate | PE/Cy7 |
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher FD reagent |
Product Type | PE-Cy7 Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction