Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Texas Red Antibody Labeling Kit
Shop All R&D Systems Products

Click to view available options
Quantity:
1 mg
30 μg
300 μg
Description
TEXAS RED is a red fluorescent dye, used in histology, in fluorescence microscopy applications and in immunohistochemistry. It has a strong absorption at 595nm, with a high fluorescence at 615nm (extinction coefficient 8.0 x10&superscript_begin;4&superscript_end;cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;) and high quantum yield.
Order Info
Quantity
Specifications
Specifications
Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
Quantity | 30 μg |
Conjugate | Texas Red |
Kit Contents | Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent |
Product Type | Rapid Texas Red Antibody Labeling Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction