Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIM Kinase 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB387849 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387849 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387849 Supplier Novus Biologicals Supplier No. NBP323404
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

LIM Kinase 1 Polyclonal antibody specifically detects LIM Kinase 1 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen LIM Kinase 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1
Host Species Rabbit
Immunogen LIM Kinase 1 (C-terminus) synthetic peptide (coupled to carrier protein) corresponding to amino acids at the C-terminus of human LIM Kinase 1. This sequence is conserved in rat and mouse LIM Kinase 1, and is not found in LIM Kinase 2. [UniProt# P53667]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3984
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.